Ball Valve, 2-Way, Straight, Parker, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
- Fittings are available in brass or stainless steel
- Treated with Siltek®/Sulfinert or Silcosteel®-CR
Parker manually, pneumatically, and electrically actuated two-way ball valves provide 1/4 turn for on-off control of fluids utilized in process and instrumentation applications.
Customers Who Bought This Also Bought