Back Ferrule, Parker, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Back ferrule of a ferrule set for providing a seal when compressed between two mating components.
- Connects to tubing
- Brass construction for corrosion resistance, ductility at high temperatures, and low magnetic permeability
Pipe and tube fittings are components used for connecting, terminating, controlling flow, and changing the direction of piping in many different industries.
Ordering information: Please note: Nuts and ferrules are not treated unless requested (custom parts). Nuts and ferrules normally are not in contact with sample pathway, and thus do not require coating.
Customers Who Bought This Also Bought