Single Taper Inlet Liner with CarboFrit® for Bruker/Varian GCs equipped with 1177 inlets, Restek
# de catalogue 75778-674
Fournisseur: Restek
Spécifications
- O.D.:6.5 mm
- Volume:0.923 ml
- Description:Single Taper Splitless, Straight with CarboFrit
- Material:Borosilicate Glass
- Cat. no.:75778-674
- For Use With:Bruker/Varian
- I.D.×L:4.0×78.5 mm
- Treatment:Siltek
- Supplier No.:21897-216.5
Spécifications
A propos de cet article
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
- Consistent quality with exceptional batch-to-batch uniformity
- Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.
Customers Who Bought This Also Bought