Uniliner® Inlet Liner with Top Taper for Bruker/Varian GCs Equipped with 1075/1077 Inlets, Restek
# de catalogue RK20346
Fournisseur: Avantor
Spécifications
- Format:Analytical
- O.D.:6.3 mm
- Description:Uniliner
- Cat. no.:RK20346
- I.D.×L:4.0×72 mm
- Supplier No.:20346
Spécifications
A propos de cet article
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
- Consistent quality with exceptional batch-to-batch uniformity
- Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.
Customers Who Bought This Also Bought