Straight/SPME Inlet Liner for Bruker/Varian GCs Equipped with 1075/1077 Inlets, Restek
# de catalogue 75779-202
Fournisseur: Restek
Spécifications
- O.D.:6.3 mm
- Volume:0.044 ml
- Description:SPME Liner
- Material:Borosilicate Glass
- Cat. no.:75779-202
- For Use With:Bruker/Varian
- I.D.×L:0.75×74 mm
- Treatment:Intermediate polarity (IP)
- Supplier No.:21113
Spécifications
A propos de cet article
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
- Consistent quality with exceptional batch-to-batch uniformity
- Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.
Customers Who Bought This Also Bought