Straight Inlet Liner with Wool for Shimadzu 17A GCs equipped with PTV inlets, Restek
# de catalogue RK21706
Fournisseur: Restek
Spécifications
- O.D.:4.0 mm
- Volume:0.191 ml
- Description:17A PTV Liner with Deactivated Wool
- Material:Borosilicate Glass
- Cat. no.:RK21706
- I.D.×L:1.6×95 mm
- Treatment:Intermediate polarity (IP)
- Supplier No.:21706
Spécifications
A propos de cet article
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
- Consistent quality with exceptional batch-to-batch uniformity
- Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.
Customers Who Bought This Also Bought