Uniliner® Inlet Liner with Wool for PerkinElmer Auto SYS™ GCs Equipped with Split/Splitless Inlet, Restek
# de catalogue RK20838
Fournisseur: Restek
Spécifications
- Description:Auto SYS Open-Top Uniliner with Deactivated Wool
- Cat. no.:RK20838
- Inner Diameter:4 mm
- For Use With:PerkinElmer® GC, Auto SYS GC
- Supplier No.:20838
Spécifications
A propos de cet article
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
- Consistent quality with exceptional batch-to-batch uniformity
- Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.
Customers Who Bought This Also Bought