Straight Inlet Liner for Agilent ¹/₄," Packed Injection Ports, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
- Consistent quality with exceptional batch-to-batch uniformity
- Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.
Customers Who Bought This Also Bought