ProFLOW 6000 Electronic Flowmeter, Restek
# de catalogue 89205-666
Fournisseur: Restek
Spécifications
- Description:Restek ProFLOW 6000 Electronic Flowmeter With Hard-Sided Carrying Case
- Cat. no.:89205-666
- Supplier No.:22656
Spécifications
A propos de cet article
With its Ex rating, the Restek ProFLOW 6000 meter is designed specifically with explosive and flammable gases in mind.
- Measure volumetric flow of most gases
- Wide range of capabilities
- Designed specifically with explosive and flammable gases in mind
Flowmeters that can measure flammable gases are becoming mandatory due to the increased use of hydrogen in chromatography. The Restek ProFLOW 6000 is the only flowmeter you need for any type of chromatography gas measurement because of its wide range of capabilities. The ProFLOW 6000 is an electronic meter capable of measuring volumetric flow for most gases. Real-time measurements can be made for various types of flow paths, including continually changing gas types. This portable unit is designed for easy handheld use, and the stand adds benchtop convenience.
Ordering information: The flowmeter is designed to measure clean, dry, non-corrosive gases. Patented.
Customers Who Bought This Also Bought