Inlet Adapter Kit for Dual Column Installation for Agilent Capillary Injectors, Split/Splitless Fitting for Capillary Columns, Restek
# de catalogue 10856-162
Fournisseur: Restek
Spécifications
- Description:Inlet Adaptor Kit for Dual Column Installation for Agilent Capillary Injectors
- Cat. no.:10856-162
- Supplier No.:27185
Spécifications
A propos de cet article
Capillary columns are easier to install due to the nut protruding farther from the insulated injection port chamber.
- 1/16-inch split/splitless fitting that accepts standard, two-hole capillary ferrules
- Same column insertion depth as the original manufacturer's equipment
For use with Agilent split/splitless fittings for capillary columns.
Kit includes adapter fitting, 1/16" capillary nut, gold-plated 1.2 mm ID dual Vespel ring inlet seal, one 0.4 mm ID two-hole ferrule, and one 0.5 mm ID two-hole ferrule.
Customers Who Bought This Also Bought