GC-MS Cleaning Kit, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Poor sensitivity, loss of sensitivity at high masses, or high multiplier gain during an auto tune are all indicators that your mass spectrometer source may need to be cleaned.
Restek has assembled all of the necessary components for cleaning and polishing your ion source.
BodyTextAccessories: This kit contains replacement cloths, micro mesh sheets, and small and large gloves for complementing your GC-MS cleaning kit.
Customers Who Bought This Also Bought