Inland 45 Pump Oil, Restek
Catalog # 89205-674
Supplier: Restek
Specifications
- Description:Inland 45 Pump Oil
- Cat. no.:89205-674
- Supplier No.:24819
Specifications
About this item
Recommended for optimum mass spec performance, this oil pump is compatible with buna-N, neoprene, and Viton® seals.
- Nontoxic
- Noncorrosive
- Ease at cold start
- Optimum vacuum pump performance
This pump oil functions at low vapor pressure 10-7 torr.
Customers Who Bought This Also Bought