EZ Twist Top® Split/Splitless Injection Port for Agilent GCs, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Gas lines are attached to the shell weldment bottom to help eliminate broken gas lines.
- Change inlet liners faster and easier
- Gas lines don't interfere with routine maintenance
- Weldment stays secure in the tool for reattachment
For use with Agilent 7890 GCs.
This weldment removal tool allows for quick removal of the hot weldment without fingers ever touching hot surfaces.
Customers Who Bought This Also Bought