Hot Swap Capillary Column Nuts, Restek
Fournisseur: Restek
Certificats
A propos de cet article
The Hot Swap capillary column nut allows you to change your capillary column while the injector temperature is still hot.
For use with "compact" Agilent-style ferrules.
Caution: For proper operation, oven fan must be kept operational during change out or risk of burn may occur.
Customers Who Bought This Also Bought