Metric Wrench Set, Restek
# de catalogue 10852-836
Fournisseur: Restek
Spécifications
- Description:Metric Wrench Set
- Cat. no.:10852-836
- Supplier No.:22997
Spécifications
A propos de cet article
High-quality 6 x 7 mm, 8 x 10 mm, and 16 x 17 mm wrenches for tightening a wide variety of fittings.
Customers Who Bought This Also Bought