Open-End Wrench Set for use with Shimadzu 17A, 2010, and 2014 Capillary Installation Gauge, Restek
# de catalogue RK22334
Fournisseur: Restek
Spécifications
- Description:Open-End Wrench Set
- Cat. no.:RK22334
- Supplier No.:22334
Spécifications
A propos de cet article
For use with Shimadzu 17A, 2010, and 2014 capillary installation gauges.
Delivery: Wrench set includes 1/4 x 5/16" and 10 x 11 mm wrenches.
Customers Who Bought This Also Bought