Liner Cap Removing Tool for Thermo Scientific GCs, Restek
Catalog # 10852-834
Supplier: Restek
Specifications
- Description:Liner Cap Removing Tool
- Cat. no.:10852-834
- Supplier No.:24937
Specifications
About this item
For use with Thermo Scientific Focus, TRACE, and TRACE Ultra GCs.
- Easily loosens the liner cap from the injector
- Unique, ergonomic handle that is easy to grip
Customers Who Bought This Also Bought