Tee Wrench, Restek
# de catalogue 10855-364
Fournisseur: Restek
Spécifications
- Description:Tee Wrench
- Cat. no.:10855-364
- Supplier No.:22623
Spécifications
A propos de cet article
Tee wrench can be used to hold 1/4" or 6 mm tee or cross fittings securely in multiple orientations during installation.
- Fits easily in toolbox, pouch, or belt
- Cushioned vinyl grip with generous gripping area
For use with Swagelok fittings only.
Customers Who Bought This Also Bought