Flexible Inspection Light
# de catalogue 10852-854
Fournisseur: Restek
Spécifications
- Description:Flexible Inspection Light
- Cat. no.:10852-854
- Supplier No.:22627
Spécifications
A propos de cet article
Use the flexible inspection light to inspect the inside surfaces of sample cylinders or other chambers.
- 14-inch reach
- 100,000-hour LED life
Customers Who Bought This Also Bought