EZ Twist Top® Split/Splitless Injection Port for Agilent GCs with Large Canister-Type Filter, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Gas lines are attached to the shell weldment bottom to help eliminate broken gas lines.
- Change inlet liners faster and easier
- Gas lines don't interfere with routine maintenance
- Weldment stays secure in the tool for reattachment
Weldment removal tool allows for quick removal of the hot weldment without fingers ever touching hot surfaces. For use with large canister-type filters.
Customers Who Bought This Also Bought