Adapters for Capillary Columns for Thermo Scientific TRACE & Focus SSL, Restek
Fournisseur: Restek
Certificats
A propos de cet article
For use with Thermo Scientific TRACE & Focus SSL and M4 ferrules.
- High-quality stainless steel construction
- Siltek®-treated version available for additional inertness
Use same installation distance as manufacturer's adapters.
Customers Who Bought This Also Bought