Helium-Specific Click-On In-Line Super Clean® Gas Trap and Connector Kits, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Helium-specific Click-On in-line Super Clean® gas trap and kits are designed specifically for purification of helium in GC-MS systems.
Delivery: Kits include ¹/₈" brass connectors and one triple trap.
Customers Who Bought This Also Bought