Click-On In-Line Super Clean® Gas Traps and Connector Kits, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Click-On fittings allow for easy, leak-tight cartridge changes, and traps are made of durable brass or stainless steel.
- High-purity output ensures 99.9999% pure gas
- Triple gas trap is ideal for purifying carrier gas
- Fuel gas trap is ideal for purifying flame ionization detector (FID) fuel gases, removing both moisture and hydrocarbons
- Helium-specific gas trap is ideal for GC-MS
Triple gas traps contain oxygen, moisture, and hydrocarbon scrubbers all in one cartridge.
Customers Who Bought This Also Bought