Capillary Installation Gauge for Thermo Scientific TRACE & Focus SSL, M4 Ferrules, Restek
# de catalogue 10852-830
Fournisseur: Restek
Spécifications
- Description:Capillary Installation Gauge
- Cat. no.:10852-830
- For Use With:for Thermo Scientific TRACE & Focus SSL (M4 ferrules)
- Supplier No.:22330
Spécifications
A propos de cet article
For use with Thermo Scientific TRACE & Focus SSL.
- Seats ferrule onto column for consistent installations
- Prevents crushed column ends
- High-quality stainless steel construction
Ordering information: For use with graphite ferrules only.
Customers Who Bought This Also Bought