FID Jet Removal Tool for Agilent 5890/6850/6890/7890 FIDs, Restek
Catalogue # 10803-284
Fournisseur: Restek
Spécifications
- Description:FID Jet Removal Tool
- Cat. no.:10803-284
- For Use With:for Agilent 5890/6850/6890/7890 GCs
- Supplier No.:22328
Spécifications
À propos de cet article
FID jet removal tools securely grip jet in socket for easy removal or installation.
- Unique, ergonomic handle that is easy to hold
- For use with Agilent 5890, 6850, 6890, and 7890 FIDs