Micropacked Inlet Conversion Kits, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Use micropacked inlet conversion kits to convert capillary GC split/splitless inlets for use with 1/16" OD micropacked columns.
- Sample pathways deactivated for ultimate inertness
For use with Agilent 5890, 6890, and 7890 GCs.
BodyTextAccessories: Replacement parts include dual Vespel ring inlet seals, and large bores (2).
Ordering information: For use with packed column FIDs only.
Customers Who Bought This Also Bought