Torx® Screwdriver Set, Restek
Catalog # 10058-844
Supplier: Restek
Specifications
- Description:Torx Screwdriver Set
- Cat. no.:10058-844
- Includes:TR-10, TR-15, and TR-20 screwdrivers
- Supplier No.:23034
Specifications
About this item
Ideal for performing routine maintenance on Agilent 6890 and 7890 GCs.
Customers Who Bought This Also Bought