ContactUsLinkComponent
Inlet Wrench for Shimadzu 17A, 2010, and 2014 GCs, Restek
Inlet Wrench for Shimadzu 17A, 2010, and 2014 GCs, Restek
Catalog # 10852-852
Supplier:  Restek
Inlet Wrench
Inlet Wrench for Shimadzu 17A, 2010, and 2014 GCs, Restek
Catalog # 10852-852
Supplier:  Restek
Specifications
  • Description:
    Inlet Wrench
  • Cat. no.:
    10852-852
  • Supplier No.:
    21159
Specifications
About this item
For use with Shimadzu 17A, 2010, and 2014 GCs.
  • Designed specifically for removing Shimadzu injection ports
  • High-quality stainless steel construction
Customers Who Bought This Also Bought