SeCure® 'Y' Connector Kits, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Combine the simplicity of a “Y” Press-Tight® connector with the strength of a metal union.
- Reliable seal integrity
- Open design allows visual confirmation of the seal
- Use to connect two analytical columns to a transfer line or guard column
The ferrules and knurled nuts hold the fused silica tubing in place, which prevents the tubing from unexpectedly disconnecting, even at temperatures as high as 400°C.
Ordering information: Kits include: SeCure® “Y” connector body, three knurled nuts, universal “Y” Press-Tight® union, and three ferrules.
Customers Who Bought This Also Bought