Graphite Ferrules for Vu2 Union® Connectors, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Graphite ferrules are designed to be suitable with 1/16" fittings.
- High-purity, high-density graphite
- Stable to 450°C
- No binders that can off-gas or adsorb analytes
- Smooth surface and clean edges
For use with Vu2 Union® connectors.
Customers Who Bought This Also Bought