Wall Mounting Bracket, Restek
# de catalogue RK21984
Fournisseur: Restek
Spécifications
- Description:Wall Mounting Bracket for Super Clean Base Plates
- Cat. no.:RK21984
- Supplier No.:21984
Spécifications
A propos de cet article
Optional wall mounting bracket allows for the mounting of Super Clean® base plates with the use of screws.
- Additional support for mounting base plates
Base plates can be mounted by using screws and the mounting holes on the base plate, or by using this optional wall mounting bracket.
Customers Who Bought This Also Bought