Replacement Needles for Agilent 7673, 7683, 7693A, and 6850 Autosampler Syringes, Hamilton, Restek
Fournisseur: Restek
Certificats
A propos de cet article
For use with Agilent 7673, 7683, 7693A, and 6850 autosamplers.
- Replacement needle volume of 5 μL–10 μL
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.