Spécifications
- Maximum Flow Rate:20 L/min
- Description:Super Clean Gas-Trapping System (2-position base plate, 2 charcoal filters)
- Max. operating pressure:11.0 bar (1100 kPa, 159 psi)
- Cat. no.:10058-716
- Fitting Size:¼"
- Supplier No.:22062
Spécifications
A propos de cet article
A Super Clean® quick-change cartridge system efficiently removes hydrocarbons from nitrogen; the Super Clean® gas trapping system purifies nitrogen and is ideal for use in LC-MS systems.
- Parallel positioning of charcoal-filled cartridges
- Efficiently removes hydrocarbons for higher gas purity
- Longer contact between nitrogen and adsorbent
The two-position base plate (installed in the gas line; standard base plate inlet/outlet fittings accept ¹/₄" tubing) allows cartridges to be exchanged without introducing impurities into the system. Spring-loaded check valves seal when a cartridge is removed and open only when a new cartridge has been locked in place. There is no need for loosening and tightening fittings every time you change cartridges, and your system cannot become contaminated during the changing process. To meet the high flow needs of the LC-MS system, the activated charcoal-filled cartridges are positioned and connected in parallel.
The incoming gas stream is split equally between the cartridges, and the two streams are rejoined after purification but before the gas exits the base plate. This approach allows longer contact between the nitrogen and the adsorbent, ensuring higher gas purity and eliminating a potential source of contaminants to your analyses. A handy date wheel, included with the system, indicates the cartridge installation date and the recommended replacement date.