Rough Pump Oil #19 for MSD Pumps, Oil Vacuum Pump, Restek
# de catalogue 10057-916
Fournisseur: Restek
Spécifications
- Description:Rough Pump Oil for MSD Pumps
- Cat. no.:10057-916
- Supplier No.:22687
Spécifications
A propos de cet article
Rough pump oil is formulated from crude oil stocks known for their durability and line-lubricating qualities.
- Long-lasting lubrication
Use in Agilent 5973/5972/5971 and GCD mass spec systems, or in other manufacturers' MSD systems that require rough pump oil.
Under average use, the oil in the foreline rough pump should be replaced every six months.
Customers Who Bought This Also Bought