Inlet Wrench for Agilent 5890/6850/6890 GCs, Restek
Catalog # 97040-462
Supplier: Restek
Specifications
- Description:Inlet Wrench
- Cat. no.:97040-462
- Supplier No.:22065
Specifications
About this item
Use an inlet wrench to remove the septum nut and weldments during GC maintenance.
- High-quality stainless steel construction
- Meets original equipment performance
For use with Agilent 5890/6850/6890 GCs.
Customers Who Bought This Also Bought