Capillary Installation Gauge for Agilent 5973/5975 MS, Restek
Catalog # 10058-700
Supplier: Restek
Specifications
- Description:Capillary Installation Gauge
- Cat. no.:10058-700
- For Use With:for Agilent 5973/5975 MS
- Supplier No.:21894
Specifications
About this item
Capillary installation gauges seat ferrules onto columns for consistent installations.
- Made from high-quality stainless steel
For use with Agilent 5973/5975 MS.
Customers Who Bought This Also Bought