PTFE Ferrules, Restek
Fournisseur: Restek
Certificats
A propos de cet article
PTFE ferrules are built to be extremely durable, with an inert nature and temperature resistance up to 250°C.
- 100% PTFE fabrication
- Completely inert
- One-piece design requires no back ferrule
- Upper temperature limit of 250°C
Customers Who Bought This Also Bought