PTFE Tip, Gas-Tight Syringes, SGE, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Interchangeable barrels, plungers, and tips extend performance and increase cost-effectiveness.
- Suitable for gases or liquids
- High accuracy of dispensed volumes
Ordering information: Needles sold separately.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.