Micro-Volume, Positive Displacement Syringes, SGE, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Sample is contained in the needle with full plunger displacement, ensuring no dead volume.
- High precision and accuracy down to 0.1 µL
- Replaceable needle/plunger kits and barrels (available on request)
Seal between the plunger and the needle can be tightened for high-pressure injection.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.