Manual Microliter Syringes, SGE, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Each syringe plunger and barrel assembly is manufactured as one working unit; components are not interchangeable or individually replaceable.
- Economical
- Available with cemented needles (N/F) or removable needles (RN/R)
Ordering information: Replacement RN/R needles available.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.