Inlet Liner Packing Tool, Restek
Catalog # 10803-214
Supplier: Restek
Specifications
- Description:Inlet Liner Packing Tool
- Cat. no.:10803-214
- Supplier No.:20339
Specifications
About this item
Recommended for inlet liners with an ID ≥2 mm.
- Accurate to a specific, measured depth
- Can be used with all manufacturers' liners
Customers Who Bought This Also Bought