Super Clean® Gas Filter Kits and Replacements, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Ensure high-purity output in these gas filters with 99.9999% pure gas (at a max flow of 2 L/min) with the use of these filter kits.
- Designed for easy, hassle-free cartridge changes
- Glass inside to prevent diffusion; polycarbonate housing outside for safety
- All traps measure 27×4.4 cm (10⁵/₈×1³/₄")
- Each base plate unit measures 10.2×10.2×4.8 cm (4×4×1⁷/₈")
Customers Who Bought This Also Bought