ECD Vent Trap, Restek
# de catalogue 10855-828
Fournisseur: Restek
Spécifications
- Description:ECD Vent Trap, 1/8"
- Cat. no.:10855-828
- Supplier No.:22017
Spécifications
A propos de cet article
The ECD vent trap reduces the release of hazardous materials from the ECD vent into the lab.
Ordering information: Includes connecting lines and mounting kit.
Customers Who Bought This Also Bought