Encapsulated Ferrules for ¹/₁₆" Compression Fittings, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Encapsulated ferrules are reusable and will not deform and stick in fittings.
- Less torque needed to seal ferrule
- Restek’s unique blend of graphite minimizes fragmentation and out-gassing
For use with 1/16-inch compression fittings.
Customers Who Bought This Also Bought