Inlet Liner Removal Tool, Restek
# de catalogue RK20181
Fournisseur: Restek
Spécifications
- Description:Inlet Liner Removal Tool
- Cat. no.:RK20181
- Supplier No.:20181
Spécifications
A propos de cet article
With this tool, easily remove liners from injectors without any more burned fingers.
- Made from high-temperature silicone
- Won’t chip or crack the liner
Customers Who Bought This Also Bought