FID Collector Assembly Kit for Agilent 6850/6890/7890 GCs, Restek
Fournisseur: Restek
Certificats
A propos de cet article
FID collector assembly kits are constructed of high-quality stainless steel and are suitable for Agilent 6850/6890/7890 GCs.
- Meets or exceeds original manufacturer's performance
- Individual replacement parts available
Also fits OI Analytical 4410 detector.
Customers Who Bought This Also Bought