Capillary Installation Gauge for Agilent GCs, Restek
Supplier: Restek
Certificates
About this item
The capillary installation gauge seats graphite ferrules onto a column for consistent installations.
- Prevents crushed column ends
- Made from high-quality stainless steel
For use with graphite ferrules only.
Ordering information: For use with graphite ferrules only.
Customers Who Bought This Also Bought