Finger-Tight Capillary Column Nuts, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Finger-tight capillary column nuts are an excellent alternative to conventional column nuts.
- Available for standard (1/16") and compact (Agilent-style) ferrules
- Made from high-quality stainless steel
They can be installed by hand without a wrench, and they cool down much faster than conventional fittings, making column and inlet maintenance easier. These stainless steel fittings work best with graphite-type ferrules and help prevent over-tightening, which can cause graphite material to exude into the inlet.
Customers Who Bought This Also Bought