Uniliner® Liner for 1/4-Inch Packed Injection Port Conversion, Restek
Fournisseur: Restek
Certificats
A propos de cet article
Uniliner® liners incorporate a gentle taper that seals the column and reduces dead volume in direct injection mode.
- Versatile—0.53 mm ID version can be used in the direct (DI) or on-column (OC) injection mode
- Available in various designs
On-column injections can be performed only with 0.53 mm ID columns because 26-gauge needles do not fit into the bore of 0.32 mm ID columns, or into the Unliner® liner taper.
Ordering information: DI = direct injection, OC = on-column injection.
Note: a Uniliner liner must be used with a Uniliner liner adaptor (cat.# 20310 or 20311) for 1/4-inch injection ports. Remember to include a liner adaptor when ordering a Uniliner® liner, unless you are purchasing replacement Uniliner® liners.
Note: a Uniliner liner must be used with a Uniliner liner adaptor (cat.# 20310 or 20311) for 1/4-inch injection ports. Remember to include a liner adaptor when ordering a Uniliner® liner, unless you are purchasing replacement Uniliner® liners.
Customers Who Bought This Also Bought