Micro-Volume, Positive Displacement Syringes, Hamilton, Restek
# de catalogue 10059-194
Fournisseur: Restek
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Volume:2 µl
- Description:Micro-Volume, Positive Displacement Syringe
- Needle Length:2.75"/70 mm
- Needle Style:2
- Needle Gauge:25
- Supplier No.:24551
- Needle Tip:KH
Spécifications
A propos de cet article
Sample is contained in the needle with full plunger displacement, ensuring no dead volume.
- High precision and accuracy down to 0.1 µL
- Replaceable needle/plunger kits and barrels (available on request)
Seal between the plunger and the needle can be tightened for high-pressure injection.
Customers Who Bought This Also Bought